Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-hydroxy-2-methylthiazole-5-carboxylate, 4-Hydroxy-2-methylthiazole-5-carboxylic acid ethyl ester, Ethyl 2-methyl-4-oxo-4,5-dihydro-1,3-thiazole-5-carboxylate

OR110310-5G
APOSOR110310-5GEA 121 EUR
APOSOR110310-5G
Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate 98%
Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate
Formel: C₇H₉NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD08275722
CAS-Nummer: 20737-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR