tert-Butyl 3-amino-5-nitro-1H-indazole-1-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110313-1G
APOSOR110313-1GEA 58 EUR
APOSOR110313-1G
tert-Butyl 3-amino-5-nitro-1H-indazole-1-carboxylate
tert-Butyl 3-amino-5-nitro-1H-indazole-1-carboxylate
Formel: C₁₂H₁₄N₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD13183010
CAS-Nummer: 574729-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR