Ethyl 4-[3-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)propoxy]benzoate

Lieferant: Apollo Scientific

OR110314-1G
APOSOR110314-1GEA 0 EUR
APOSOR110314-1G
Ethyl 4-[3-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)propoxy]benzoate
Ethyl 4-[3-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)propoxy]benzoate
Formel: C₂₀H₁₉NO₅
Molecular Weight: 353,369 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06205031
CAS-Nummer: 100840-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR