(2-Phenyl-1,3-thiazol-5-yl)methanol

Lieferant: Apollo Scientific

OR110315-1G
APOSOR110315-1GEA 0 EUR
APOSOR110315-1G
(2-Phenyl-1,3-thiazol-5-yl)methanol
(2-Phenyl-1,3-thiazol-5-yl)methanol
Formel: C₁₀H₉NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD09694149
CAS-Nummer: 859485-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR