4-(2,4-Dichlorophenyl)-1,3-thiazol-2(3H)-one

Lieferant: Apollo Scientific

OR110317-1G
APOSOR110317-1GEA 0 EUR
APOSOR110317-1G
4-(2,4-Dichlorophenyl)-1,3-thiazol-2(3H)-one
4-(2,4-Dichlorophenyl)-1,3-thiazol-2(3H)-one
Formel: C₉H₅Cl₂NOS
Molecular Weight: 246,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09746489
CAS-Nummer: 42489-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR