2-Amino-5-chloro-4-methylnicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR110320-1G
APOSOR110320-1GEA 284 EUR
APOSOR110320-1G
2-Amino-5-chloro-4-methylnicotinonitrile
2-Amino-5-chloro-4-methylnicotinonitrile
Formel: C₇H₆ClN₃
Molecular Weight: 167,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583149
CAS-Nummer: 1305712-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR