3-Brom-4-nitro-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR110321-1G
APOSOR110321-1GEA
282
EUR
APOSOR110321-1G
3-Brom-4-nitro-1H-indazol
3-Bromo-4-nitro-1H-indazole
Formel:
C₇H₄BrN₃O₂ Schmelzpunkt: 214…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781572 CAS-Nummer: 74209-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...