3-Chloro-4-nitro-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR110322-1G
APOSOR110322-1GEA 418 EUR
APOSOR110322-1G
3-Chloro-4-nitro-1H-indazole
3-Chloro-4-nitro-1H-indazole
Formel: C₇H₄ClN₃O₂
Molecular Weight: 197,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263201
CAS-Nummer: 54768-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR