4-Methyl-6-(1,3-thiazol-2-yl)pyrimidin-2-amine

Lieferant: Apollo Scientific

OR110324-1G
APOSOR110324-1GEA 0 EUR
APOSOR110324-1G
4-Methyl-6-(1,3-thiazol-2-yl)pyrimidin-2-amine
4-Methyl-6-(1,3-thiazol-2-yl)pyrimidin-2-amine
Formel: C₈H₈N₄S
Molecular Weight: 192,241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705823
CAS-Nummer: 913322-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR