2-(1-Methylhydrazino)-5-nitropyridine

Lieferant: Apollo Scientific
Achtung

OR110329-1G
APOSOR110329-1GEA 58 EUR
APOSOR110329-1G
2-(1-Methylhydrazino)-5-nitropyridine
2-(1-Methylhydrazino)-5-nitropyridine
Formel: C₆H₈N₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10000050
CAS-Nummer: 41286-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR