2-Propyl[1,3]oxazolo[4,5-b]pyridine

Lieferant: Apollo Scientific

OR110331-1G
APOSOR110331-1GEA 0 EUR
APOSOR110331-1G
2-Propyl[1,3]oxazolo[4,5-b]pyridine
2-Propyl[1,3]oxazolo[4,5-b]pyridine
Formel: C₉H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07021391
CAS-Nummer: 104711-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR