2-Methyl-7-nitro-2H-indazole

Lieferant: Apollo Scientific

OR110332-1G
APOSOR110332-1GEA 0 EUR
APOSOR110332-1G
2-Methyl-7-nitro-2H-indazole
2-Methyl-7-nitro-2H-indazole
Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01719664
CAS-Nummer: 13436-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR