1-Methyl-7-nitro-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR110333-1G
APOSOR110333-1GEA 87 EUR
APOSOR110333-1G
1-Methyl-7-nitro-1H-indazole
1-Methyl-7-nitro-1H-indazole
Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832095
CAS-Nummer: 58706-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR