2-Butyl-1,3-benzothiazole

Lieferant: Apollo Scientific

OR110337-1G
APOSOR110337-1GEA 0 EUR
APOSOR110337-1G
2-Butyl-1,3-benzothiazole
2-Butyl-1,3-benzothiazole
Formel: C₁₁H₁₃NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00972072
CAS-Nummer: 54798-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR