2-Propyl-1,3-benzothiazole

Lieferant: Apollo Scientific
Achtung

OR110338-1G
APOSOR110338-1GEA 109 EUR
APOSOR110338-1G
2-Propyl-1,3-benzothiazole
2-Propyl-1,3-benzothiazole
Formel: C₁₀H₁₁NS
Lagertemperatur: Raumtemperatur
MDL: MFCD15146801
CAS-Nummer: 17229-76-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR