2-Ethyl-1,3-benzothiazole

Lieferant: Apollo Scientific

OR110339-1G
APOSOR110339-1GEA 0 EUR
APOSOR110339-1G
2-Ethyl-1,3-benzothiazole
2-Ethyl-1,3-benzothiazole
Formel: C₉H₉NS
Molecular Weight: 163,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05982141
CAS-Nummer: 936-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR