5-Chloro-2-methyl-6-nitro-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-2-methyl-6-nitrobenzo[d]oxazole

OR110340-1G
APOSOR110340-1GEA 21.9 EUR
APOSOR110340-1G
5-Chloro-2-methyl-6-nitro-1,3-benzoxazole
5-Chloro-2-methyl-6-nitro-1,3-benzoxazole
Formel: C₈H₅ClN₂O₃
Molecular Weight: 212,59 g/mol
Schmelzpunkt: 147…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00804971
CAS-Nummer: 13452-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR