5-Chloro-2-ethyl-6-nitro-1,3-benzoxazole

Lieferant: Apollo Scientific

OR110342-1G
APOSOR110342-1GEA 0 EUR
APOSOR110342-1G
5-Chloro-2-ethyl-6-nitro-1,3-benzoxazole
5-Chloro-2-ethyl-6-nitro-1,3-benzoxazole
Formel: C₉H₇ClN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05663829
CAS-Nummer: 886361-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR