2-(Ethylamino)nicotinonitril

Lieferant: Apollo Scientific

OR110343-5G
APOSOR110343-5GEA 109 EUR
APOSOR110343-5G
2-(Ethylamino)nicotinonitril
2-(Ethylamino)nicotinonitril
Formel: C₈H₉N₃
Molecular Weight: 147,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09935524
CAS-Nummer: 52583-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR