2-(2-Methyl-1,3-thiazol-4-yl)acetohydrazide

Lieferant: Apollo Scientific
Achtung

OR110344-1G
APOSOR110344-1GEA 116 EUR
APOSOR110344-1G
2-(2-Methyl-1,3-thiazol-4-yl)acetohydrazide
2-(2-Methyl-1,3-thiazol-4-yl)acetohydrazide
Formel: C₆H₉N₃OS
Lagertemperatur: Raumtemperatur
MDL: MFCD03789239
CAS-Nummer: 496057-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR