5-Chloro-2-(methylamino)nicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR110346-1G
APOSOR110346-1GEA 342 EUR
APOSOR110346-1G
5-Chloro-2-(methylamino)nicotinonitrile
5-Chloro-2-(methylamino)nicotinonitrile
Formel: C₇H₆ClN₃
Molecular Weight: 167,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15146802
CAS-Nummer: 1427460-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR