5-Bromo-2-(methylamino)nicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR110347-1G
APOSOR110347-1GEA 203 EUR
APOSOR110347-1G
5-Bromo-2-(methylamino)nicotinonitrile
5-Bromo-2-(methylamino)nicotinonitrile
Formel: C₇H₆BrN₃
Molecular Weight: 212,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619580
CAS-Nummer: 1346534-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR