2,5-Dibromo-1H-imidazol-4-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR110349-1G
APOSOR110349-1GEA
319
EUR
APOSOR110349-1G
2,5-Dibromo-1H-imidazol-4-amine
2,5-Dibromo-1H-imidazol-4-amine
Formel:
C₃H₃Br₂N₃ Molecular Weight: 240,884 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10001383 CAS-Nummer: 1029720-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...