2-Acetyl-7-bromo-5-chloro-2H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
OR110361-1G
APOSOR110361-1GEA
322
EUR
APOSOR110361-1G
2-Acetyl-7-bromo-5-chloro-2H-indazole
2-Acetyl-7-bromo-5-chloro-2H-indazole
Formel:
C₉H₆BrClN₂O Molecular Weight: 273,514 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16619589 CAS-Nummer: 1355171-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...