Methyl-4-methyl-3-nitrobenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-methyl-3-nitrobenzoate
OR110363-25G
APOSOR110363-25GEA
21.9
EUR
APOSOR110363-25G
Methyl-4-methyl-3-nitrobenzoat
Methyl-4-methyl-3-nitrobenzoat
Formel:
C₉H₉NO₄ Molecular Weight: 195.18 g/mol Schmelzpunkt: 49…53 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130054 CAS-Nummer: 7356-11-8 EINECS: 230-886-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...