Ethyl 4-amino-3-bromo-5-methylbenzoate

Lieferant: Apollo Scientific
Achtung

OR110365-1G
APOSOR110365-1GEA 140 EUR
APOSOR110365-1G
Ethyl 4-amino-3-bromo-5-methylbenzoate
Ethyl 4-amino-3-bromo-5-methylbenzoate
Formel: C₁₀H₁₂BrNO₂
Molecular Weight: 258,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16631259
CAS-Nummer: 1379303-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR