2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR110370-1G
APOSOR110370-1GEA 557 EUR
APOSOR110370-1G
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid
Formel: C₁₀H₉NO₄
Molecular Weight: 207,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16631264
CAS-Nummer: 1195159-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR