Quinoxaline-6-carboxamide

Lieferant: Apollo Scientific
Achtung

OR110372-1G OR110372-250MG
APOSOR110372-1GEA 142 EUR
APOSOR110372-1G APOSOR110372-250MG
Quinoxaline-6-carboxamide
Quinoxaline-6-carboxamide
Formel: C₉H₇N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09955631
CAS-Nummer: 457882-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR