N-Methylquinoxaline-6-carboxamide

Lieferant: Apollo Scientific
Achtung

OR110373-1G
APOSOR110373-1GEA 293 EUR
APOSOR110373-1G
N-Methylquinoxaline-6-carboxamide
N-Methylquinoxaline-6-carboxamide
Formel: C₁₀H₉N₃O
Molecular Weight: 187.198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16631267
CAS-Nummer: 1343299-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR