1H-Indazole-6-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR110374-1G
APOSOR110374-1GEA
207
EUR
APOSOR110374-1G
1H-Indazole-6-carbohydrazide
1H-Indazole-6-carbohydrazide
Formel:
C₈H₈N₄O Molecular Weight: 176,175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11506091 CAS-Nummer: 1086392-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...