1-Boc-4-(4-nitrophenyl)piperazine 95%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-(4-nitrophenyl)piperazine-1-carboxylate, 1-Boc-4-(4-Nitrophenyl)piperazine

OR110377-5G
APOSOR110377-5GEA 69.6 EUR
APOSOR110377-5G
1-Boc-4-(4-nitrophenyl)piperazine 95%
1-Boc-4-(4-nitrophenyl)piperazine
Formel: C₁₅H₂₁N₃O₄
Molecular Weight: 307,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187326
CAS-Nummer: 182618-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR