2-Amino-5-chlor-3-iodpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chlor-3-iodpyridin-2-amin, 5-Chlor-3-iod-2-pyridylamin

OR110378-25G
APOSOR110378-25GEA 116 EUR
APOSOR110378-25G
2-Amino-5-chlor-3-iodpyridin
2-Amino-5-chlor-3-iodpyridin
Formel: C₅H₄ClIN₂
Molecular Weight: 254.46 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07644578
CAS-Nummer: 211308-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR