1H-Indole-3-carbohydrazide

Lieferant: Apollo Scientific

OR110382-1G
APOSOR110382-1GEA 94.4 EUR
APOSOR110382-1G
1H-Indole-3-carbohydrazide
1H-Indole-3-carbohydrazide
Formel: C₉H₉N₃O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 15317-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR