Ethyl 7-bromo-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110383-1G
APOSOR110383-1GEA 0 EUR
APOSOR110383-1G
Ethyl 7-bromo-1-benzothiophene-2-carboxylate
Ethyl 7-bromo-1-benzothiophene-2-carboxylate
Formel: C₁₁H₉BrO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD17168278
CAS-Nummer: 1355171-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR