Ethyl 4-amino-3-nitrobenzoate

Lieferant: Apollo Scientific

OR110387-1G
APOSOR110387-1GEA 0 EUR
APOSOR110387-1G
Ethyl 4-amino-3-nitrobenzoate
Ethyl 4-amino-3-nitrobenzoate
Formel: C₉H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00044015
CAS-Nummer: 76918-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR