2-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific
Achtung

OR110388-1G
APOSOR110388-1GEA 372 EUR
APOSOR110388-1G
2-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
2-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
Formel: C₉H₈N₂O₄
Molecular Weight: 208,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17168279
CAS-Nummer: 132522-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR