Ethyl 6-amino-5-chloronicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-5-chloropyridine-3-carboxylic acid ethyl ester

OR110392-1G
APOSOR110392-1GEA 92.8 EUR
APOSOR110392-1G
Ethyl 6-amino-5-chloronicotinate
Ethyl 6-amino-5-chloronicotinate
Formel: C₈H₉ClN₂O₂
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD15523642
CAS-Nummer: 305329-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR