Ethyl 4-amino-3-bromo-5-nitrobenzoate

Lieferant: Apollo Scientific

OR110397-1G
APOSOR110397-1GEA 74.1 EUR
APOSOR110397-1G
Ethyl 4-amino-3-bromo-5-nitrobenzoate
Ethyl 4-amino-3-bromo-5-nitrobenzoate
Formel: C₉H₉BrN₂O₄
Molecular Weight: 289,083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044014
CAS-Nummer: 82760-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR