N-(3-Nitrophenyl)hydrazinecarboxamide

Lieferant: Apollo Scientific
Achtung

OR110399-1G
APOSOR110399-1GEA 58 EUR
APOSOR110399-1G
N-(3-Nitrophenyl)hydrazinecarboxamide
N-(3-Nitrophenyl)hydrazinecarboxamide
Formel: C₇H₈N₄O₃
Molecular Weight: 196,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16170908
CAS-Nummer: 17433-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR