N-(1-Methyl-1H-indazol-7-yl)thiourea

Lieferant: Apollo Scientific
Achtung

OR110401-1G
APOSOR110401-1GEA 580 EUR
APOSOR110401-1G
N-(1-Methyl-1H-indazol-7-yl)thiourea
N-(1-Methyl-1H-indazol-7-yl)thiourea
Formel: C₉H₁₀N₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD17168291
CAS-Nummer: 41968-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR