1,3,5-Trimethyl-1H-pyrazole-4-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110408-1G
APOSOR110408-1GEA 180 EUR
APOSOR110408-1G
1,3,5-Trimethyl-1H-pyrazole-4-carbohydrazide
1,3,5-Trimethyl-1H-pyrazole-4-carbohydrazide
Formel: C₇H₁₂N₄O
Molecular Weight: 168,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02186021
CAS-Nummer: 477711-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR