7-Methoxy-1H-indazole 95%

Lieferant: Apollo Scientific
Achtung

OR110413-1G
APOSOR110413-1GEA 217 EUR
APOSOR110413-1G
7-Methoxy-1H-indazole 95%
7-Methoxy-1H-indazole
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08752584
CAS-Nummer: 133841-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR