4-(Hydrazinocarbonyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR110418-1G OR110418-250MG
APOSOR110418-1GEA 168 EUR
APOSOR110418-1G APOSOR110418-250MG
4-(Hydrazinocarbonyl)benzoic acid
4-(Hydrazinocarbonyl)benzoic acid
Formel: C₈H₈N₂O₃
Molecular Weight: 180.161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041381
CAS-Nummer: 46206-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR