4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one

Lieferant: Apollo Scientific
Achtung

OR110421-1G OR110421-5G OR110421-10G
APOSOR110421-1GEA 115 EUR
APOSOR110421-1G APOSOR110421-5G APOSOR110421-10G
4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Formel: C₈H₆BrN₃O
Molecular Weight: 240.057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384918
CAS-Nummer: 1007578-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR