6-Bromo-2,3-dimethylquinoxaline

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-2,3-dimethyl-1,4-benzodiazine

OR110426-1G
APOSOR110426-1GEA 0 EUR
APOSOR110426-1G
6-Bromo-2,3-dimethylquinoxaline
6-Bromo-2,3-dimethylquinoxaline
Formel: C₁₀H₉BrN₂
Molecular Weight: 237,096 g/mol
Schmelzpunkt: 66…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11111652
CAS-Nummer: 18470-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR