6-Bromoimidazo[1,2-a]pyridine-2-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR110428-1G
APOSOR110428-1GEA
140
EUR
APOSOR110428-1G
6-Bromoimidazo[1,2-a]pyridine-2-carbohydrazide
6-Bromoimidazo[1,2-a]pyridine-2-carbohydrazide
Formel:
C₈H₇BrN₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD18804151 CAS-Nummer: 474956-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...