1H-Benzimidazole-5-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110435-1G
APOSOR110435-1GEA 232 EUR
APOSOR110435-1G
1H-Benzimidazole-5-carbohydrazide
1H-Benzimidazole-5-carbohydrazide
Formel: C₈H₈N₄O
Molecular Weight: 176,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703627
CAS-Nummer: 108038-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR