5-(4-Iodophenyl)-1,3-oxazole

Lieferant: Apollo Scientific
Achtung

OR110436-1G
APOSOR110436-1GEA 186 EUR
APOSOR110436-1G
5-(4-Iodophenyl)-1,3-oxazole
5-(4-Iodophenyl)-1,3-oxazole
Formel: C₉H₆INO
Molecular Weight: 271,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024923
CAS-Nummer: 1206169-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR