3-Bromo-8-nitroimidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR110437-1G
APOSOR110437-1GEA
186
EUR
APOSOR110437-1G
3-Bromo-8-nitroimidazo[1,2-a]pyridine
3-Bromo-8-nitroimidazo[1,2-a]pyridine
Formel:
C₇H₄BrN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17016106 CAS-Nummer: 52310-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...