1-Methyl-1H-indole-5-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR110439-1G
APOSOR110439-1GEA 48 EUR
APOSOR110439-1G
1-Methyl-1H-indole-5-carbonitrile
1-Methyl-1H-indole-5-carbonitrile
Formel: C₁₀H₈N₂
Molecular Weight: 156.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025840
CAS-Nummer: 91634-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR